Learn More
(Benzoylmethylene)triphenylphosphorane, 98+%, Thermo Scientific Chemicals
$80.38 - $850.60
Chemical Identifiers
CAS | 859-65-4 |
---|---|
Molecular Formula | C26H21OP |
Molecular Weight (g/mol) | 380.43 |
MDL Number | MFCD00014088 |
InChI Key | MZRSAJZDYIISJW-UHFFFAOYSA-N |
Synonym | benzoylmethylene triphenylphosphorane, 2-triphenylphosphoranylidene acetophenone, 1-phenyl-2-triphenylphosphoranylidene ethanone, phenacylidenetriphenyl phosphorane, phenylcarbonylmethylene triphenylphosphorane, acetophenone, 2-triphenylphosphoranylidene, phenacylidenetriphenylphosphorane, triphenylphosphinebenzoylmethylene, ethanone, 1-phenyl-2-triphenylphosphoranylidene |
PubChem CID | 70073 |
IUPAC Name | 1-phenyl-2-(triphenyl-$l^{5}-phosphanylidene)ethanone |
SMILES | O=C(C=P(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=CC=C1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1218406
|
Thermo Scientific Chemicals
A1218406 |
5 g |
N/A
|
|
|||||
AAA1218414
|
Thermo Scientific Chemicals
A1218414 |
25 g |
N/A
|
|
|||||
AAA1218422
|
Thermo Scientific Chemicals
A1218422 |
100 g |
N/A
|
|
|||||
Description
It is used in organic synthesis, wittig reagent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is used in organic synthesis, wittig reagent.
Solubility
Soluble in Methanol (almost transparency).
Notes
Air sensitive. Store in refrigerator. Store away from oxidizing agent, air.
Chemical Identifiers
859-65-4 | |
380.43 | |
MZRSAJZDYIISJW-UHFFFAOYSA-N | |
70073 | |
O=C(C=P(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=CC=C1 |
C26H21OP | |
MFCD00014088 | |
benzoylmethylene triphenylphosphorane, 2-triphenylphosphoranylidene acetophenone, 1-phenyl-2-triphenylphosphoranylidene ethanone, phenacylidenetriphenyl phosphorane, phenylcarbonylmethylene triphenylphosphorane, acetophenone, 2-triphenylphosphoranylidene, phenacylidenetriphenylphosphorane, triphenylphosphinebenzoylmethylene, ethanone, 1-phenyl-2-triphenylphosphoranylidene | |
1-phenyl-2-(triphenyl-$l^{5}-phosphanylidene)ethanone |
Specifications
859-65-4 | |
C26H21OP | |
5 g | |
Air sensitive | |
Soluble in Methanol (almost transparency). | |
O=C(C=P(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=CC=C1 | |
380.43 | |
380.43 | |
(Benzoylmethylene)triphenylphosphorane |
183°C to 186°C | |
MFCD00014088 | |
620285 | |
benzoylmethylene triphenylphosphorane, 2-triphenylphosphoranylidene acetophenone, 1-phenyl-2-triphenylphosphoranylidene ethanone, phenacylidenetriphenyl phosphorane, phenylcarbonylmethylene triphenylphosphorane, acetophenone, 2-triphenylphosphoranylidene, phenacylidenetriphenylphosphorane, triphenylphosphinebenzoylmethylene, ethanone, 1-phenyl-2-triphenylphosphoranylidene | |
MZRSAJZDYIISJW-UHFFFAOYSA-N | |
1-phenyl-2-(triphenyl-$l^{5}-phosphanylidene)ethanone | |
70073 | |
≥98% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 212-727-2
RTECSNumber : AN0540000
TSCA : No
Recommended Storage : Keep cold
RUO – Research Use Only