missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Benzoylformic acid, 97%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 105760250
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Benzoylformic acid | |
Authentic | |
Glass bottle | |
C6H5COCO2H | |
25g | |
benzoylformic acid, phenylglyoxylic acid, oxophenylacetic acid, benzeneglyoxylic acid, phenylgloxylic acid, 2-phenylethanoic acid, oxo phenyl acetic acid, benzoylformate, phenylglyoxalate, phenyloxoacetic acid | |
FAQJJMHZNSSFSM-UHFFFAOYSA-N | |
2-oxo-2-phenylacetic acid | |
11915 | |
150.13 |
611-73-4 | |
97% | |
C8H6O3 | |
MFCD00002575 | |
10,654 | |
Solubility in water: .. Other solubilities: 1g/10mL methanol | |
OC(=O)C(=O)C1=CC=CC=C1 | |
150.13 | |
CHEBI:18280 | |
97% |
Safety and Handling
GHS Signal Word: Warning
EINECSNumber : 210-278-7
RUO â Research Use Only