missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Benzoylferrocene 98.0+%, TCI America™
$104.52 - $267.10
Chemical Identifiers
CAS | 1272-44-2 |
---|---|
Molecular Formula | C17H14FeO |
Molecular Weight (g/mol) | 290.14 |
MDL Number | MFCD00001431 |
InChI Key | JJHHJZCYDPLHIL-UHFFFAOYNA-N |
IUPAC Name | Benzoylferrocene |
SMILES | [Fe].c1cccc1.O=C(c1cccc1)C1=CC=CC=C1 |
Chemical Identifiers
1272-44-2 | |
290.14 | |
JJHHJZCYDPLHIL-UHFFFAOYNA-N | |
[Fe].c1cccc1.O=C(c1cccc1)C1=CC=CC=C1 |
C17H14FeO | |
MFCD00001431 | |
Benzoylferrocene |
Specifications
1272-44-2 | |
C17H14FeO | |
5 g | |
[Fe].c1cccc1.O=C(c1cccc1)C1=CC=CC=C1 | |
290.14 | |
≥98.0% (GC) | |
Benzoylferrocene |
110°C | |
MFCD00001431 | |
JJHHJZCYDPLHIL-UHFFFAOYNA-N | |
Benzoylferrocene | |
290.14 | |
Crystalline Powder |
Safety and Handling
TSCA : Yes