Please login to your online account to display your discounted pricing

Alfa Aesar™ Benzoylferrocene, 98%

$31.14 - $31.14

Chemical Identifiers

Chemical Name or Material Benzoylferrocene
CAS 1272-44-2
Molecular Formula C17H14FeO
Formula Weight 290.14g/mol
MDL Number MFCD00001431
Synonym Ferrocenyl phenyl ketone
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1768503

 
alfa aesar™
A17685-03
1g Each for $31.14
Description & Specifications

Specifications

Melting Point 106° to 109°C
Quantity 1g
Chemical Name or Material Benzoylferrocene
CAS 1272-44-2
Assay 98%
Molecular Formula C17H14FeO
Formula Weight 290.14g/mol
MDL Number MFCD00001431
Synonym Ferrocenyl phenyl ketone
TSCA TSCA