Please login to your online account to display your discounted pricing

Benzoyleneurea, 98%, ACROS Organics™


Quantity 100g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC161771000

 View more versions of this product

Catalog No. AC161771000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Fluffy Powder
Formula Weight 162.15
Assay ≥97.5%
Quantity 100g
Color White to light pink
Beilstein 24, 373
CAS Max % 100.0
Packaging Glass bottle
Infrared Spectrum Authentic
Assay Percent Range 98%
MDL Number MFCD00006699
CAS Min % 97.5
Synonym 2, 4(1H, 3H)-Quinazolinedione
CAS 86-96-4
Chemical Name or Material Benzoyleneurea
Molecular Formula C8H6N2O2
Name Note 98%

Specifications

Chemical Name or Material Benzoyleneurea
Name Note 98%
CAS 86-96-4
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C8H6N2O2
Formula Weight 162.15
MDL Number MFCD00006699
Synonym 2, 4(1H, 3H)-Quinazolinedione