missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Benzoyleneurea, 98%, ACROS Organics™

Chemical Identifiers

CAS 86-96-4
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Molecular Formula C8H6N2O2
Formula Weight 162.15
MDL Number MFCD00006699
Synonym 2, 4(1H, 3H)-Quinazolinedione
Chemical Name or Material Benzoyleneurea, 98%
Assay ≥97.5%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC161771000 SDS Thermo Fisher Scientific
AC161771000
Glass bottle 100g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

86-96-4
100.0
C8H6N2O2
MFCD00006699
Benzoyleneurea, 98%
97.5
98%
162.15
2, 4(1H, 3H)-Quinazolinedione
≥97.5%

Specifications

86-96-4
100.0
98%
162.15
24, 373
2, 4(1H, 3H)-Quinazolinedione
White to light pink
Benzoyleneurea, 98%
97.5
Fluffy Powder
C8H6N2O2
MFCD00006699
Authentic
Glass bottle
100g
≥97.5%