Please login to your online account to display your discounted pricing

Alfa Aesar™ Benzoylcholine iodide, 98+%

$42.19 - $42.19

Chemical Identifiers

Chemical Name or Material Benzoylcholine iodide
CAS 17518-43-3
Molecular Formula C12H18INO2
Linear Formula C6H5CO2CH2CH2N(CH3)3I
Formula Weight 335.19
MDL Number MFCD00038726
Beilstein 3576708
Synonym (2-Benzoyloxyethyl)trimethylammonium iodide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2238703

 
alfa aesar™
B2238703
1g Each for $42.19
Description & Specifications

Specifications

Formula Weight 335.19
Chemical Name or Material Benzoylcholine iodide
TSCA TSCA
Assay ≥98%
Molecular Formula C12H18INO2
Linear Formula C6H5CO2CH2CH2N(CH3)3I
MDL Number MFCD00038726
CAS 17518-43-3
Beilstein 3576708
Synonym (2-Benzoyloxyethyl)trimethylammonium iodide
Melting Point ≈245°C (dec.)
Quantity 1g
Recommended Storage 2° to 8°C
Safety and Handling

2° to 8°C