Please login to your online account to display your discounted pricing

Alfa Aesar™ Benzoylcholine chloride, 99%

$41.77 - $565.76

Chemical Identifiers

Chemical Name or Material Benzoylcholine chloride
CAS 2964-09-2
Molecular Formula C6H5CO2CH2CH2N(CH3)3Cl
Formula Weight 243.73g/mol
MDL Number MFCD00011786
Beilstein 3919727
Synonym (2-Benzoyloxyethyl)trimethylammonium chloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1517909

 
alfa aesar™
A15179-09
10g Each for $41.77

AAA1517918

 
alfa aesar™
A15179-18
50g Each for $176.64

AAA1517930

 
alfa aesar™
A15179-30
250g Each for $565.76
Description & Specifications

Specifications

Melting Point 205° to 208°C
Quantity 10g
Chemical Name or Material Benzoylcholine chloride
CAS 2964-09-2
Assay 99%
Molecular Formula C6H5CO2CH2CH2N(CH3)3Cl
Formula Weight 243.73g/mol
MDL Number MFCD00011786
Beilstein 3919727
Synonym (2-Benzoyloxyethyl)trimethylammonium chloride
RTECS Number GA0830000