missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Benzoylacetonitrile, 98+%, ACROS Organics™

Chemical Identifiers

CAS 614-16-4
Molecular Formula C9H7NO
Molecular Weight (g/mol) 145.16
MDL Number MFCD00001942
InChI Key ZJRCIQAMTAINCB-UHFFFAOYSA-N
Synonym 2-cyanoacetophenone, 3-oxo-3-phenyl-propionitrile, 3-phenyl-3-ketopropionitrile, acetonitrile, benzoyl, benzenepropanenitrile, beta-oxo, benzoyl acetonitrile, benzoylacetonitrile, cyanoacetophenone, cyanomethyl phenyl ketone, phenacyl cyanide
PubChem CID 64799
ChEBI CHEBI:51855
IUPAC Name 3-oxo-3-phenylpropanenitrile
SMILES C1=CC=C(C=C1)C(=O)CC#N
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC402000050
SDS View Documents
Thermo Fisher Scientific
402000050
5g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

614-16-4
145.16
ZJRCIQAMTAINCB-UHFFFAOYSA-N
64799
3-oxo-3-phenylpropanenitrile
C9H7NO
MFCD00001942
2-cyanoacetophenone, 3-oxo-3-phenyl-propionitrile, 3-phenyl-3-ketopropionitrile, acetonitrile, benzoyl, benzenepropanenitrile, beta-oxo, benzoyl acetonitrile, benzoylacetonitrile, cyanoacetophenone, cyanomethyl phenyl ketone, phenacyl cyanide
CHEBI:51855
C1=CC=C(C=C1)C(=O)CC#N

Specifications

Glass bottle
78.0°C to 82.0°C
614-16-4
100.0
C9H7NO
MFCD00001942
2-cyanoacetophenone, 3-oxo-3-phenyl-propionitrile, 3-phenyl-3-ketopropionitrile, acetonitrile, benzoyl, benzenepropanenitrile, beta-oxo, benzoyl acetonitrile, benzoylacetonitrile, cyanoacetophenone, cyanomethyl phenyl ketone, phenacyl cyanide
ZJRCIQAMTAINCB-UHFFFAOYSA-N
3-oxo-3-phenylpropanenitrile
64799
145.16
Benzoylacetonitrile, ≥98%
160.0°C (10.0 mmHg)
5g
98.0
98+%
C6H5COCH2CN
10, 680
Solubility in water: slightly soluble
C1=CC=C(C=C1)C(=O)CC#N
145.16
CHEBI:51855
98+%
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Harmful in contact with skin.
Harmful if inhaled.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor

GHS Signal Word: Warning

EINECSNumber : 210-369-1

TSCA : TSCA

SDS