Please login to your online account to display your discounted pricing

Alfa Aesar™ Benzoyl-Phe-Ala-Arg

For cellular and molecular biology applications

Specifications

Product Type Benzoyl-Phe-Ala-Arg
Molecular Weight 496.56
Storage Requirements -30° to -10°C
Sequence Benzoyl-Phe-Ala-Arg
Format Solid
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ66075LB0

 
alfa aesar™
J66075LB0
5mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ66075MCR

 
alfa aesar™
J66075MCR
1mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Product Type Benzoyl-Phe-Ala-Arg
Molecular Weight 496.56
Storage Requirements -30° to -10°C
Sequence Benzoyl-Phe-Ala-Arg
Format Solid
Regulatory Status RUO
Purification Method HPLC