missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Benzoyl-L-histidine Monohydrate, TCI America™

$30.56 - $53.47

Chemical Identifiers

CAS 5354-94-9
Molecular Formula C13H13N3O3
Molecular Weight (g/mol) 259.265
MDL Number MFCD00037850
InChI Key AUDPUFBIVWMAED-NSHDSACASA-N
PubChem CID 152263
IUPAC Name (2S)-2-benzamido-3-(1H-imidazol-5-yl)propanoic acid
SMILES C1=CC=C(C=C1)C(=O)NC(CC2=CN=CN2)C(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B02041G View Documents TCI America
B02041G
1g Each for $53.47
 
B0204100MG View Documents TCI America
B0204100MG
100mg Each for $30.56
 
Specifications

Chemical Identifiers

5354-94-9
259.265
AUDPUFBIVWMAED-NSHDSACASA-N
(2S)-2-benzamido-3-(1H-imidazol-5-yl)propanoic acid
C13H13N3O3
MFCD00037850
152263
C1=CC=C(C=C1)C(=O)NC(CC2=CN=CN2)C(=O)O

Specifications

White
5354-94-9
MFCD00037850
C1=CC=C(C=C1)C(=O)NC(CC2=CN=CN2)C(=O)O
259.265
259.27
≥98.0% (HPLC,T)
1g
C13H13N3O3
AUDPUFBIVWMAED-NSHDSACASA-N
(2S)-2-benzamido-3-(1H-imidazol-5-yl)propanoic acid
152263
Crystal-Powder at 20°C
Benzoyl-L-histidine Monohydrate
Safety and Handling

Safety and Handling

TSCA : No