Learn More
Benzoyl isothiocyanate, 98%, Thermo Scientific Chemicals
$85.07 - $85.07
Chemical Identifiers
CAS | 532-55-8 |
---|---|
Molecular Formula | C8H5NOS |
Molecular Weight (g/mol) | 163.2 |
MDL Number | MFCD00004815 |
InChI Key | CPEKAXYCDKETEN-UHFFFAOYSA-N |
Synonym | benzoylthiocarbimide, benzoylisothiocyanate, benzoylisothiocyante, isothiocyanic acid benzoyl ester, unii-u8j7q32b86, n-benzoyl isothiocyanate, benzoic acid, anhydride with hncs, benzoic acid, anhydride with isothiocyanic acid, oxophenylmethanisothiocyanate, 1-benzoyl acetone |
PubChem CID | 68284 |
IUPAC Name | benzoyl isothiocyanate |
SMILES | C1=CC=C(C=C1)C(=O)N=C=S |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC314630050
|
Thermo Scientific Chemicals
314630050 |
5 g | Glass bottle |
Each for $85.07
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
532-55-8 | |
163.2 | |
CPEKAXYCDKETEN-UHFFFAOYSA-N | |
68284 | |
C1=CC=C(C=C1)C(=O)N=C=S |
C8H5NOS | |
MFCD00004815 | |
benzoylthiocarbimide, benzoylisothiocyanate, benzoylisothiocyante, isothiocyanic acid benzoyl ester, unii-u8j7q32b86, n-benzoyl isothiocyanate, benzoic acid, anhydride with hncs, benzoic acid, anhydride with isothiocyanic acid, oxophenylmethanisothiocyanate, 1-benzoyl acetone | |
benzoyl isothiocyanate |
Specifications
532-55-8 | |
100.0 | |
128.0°C to 131.0°C (15.0 mmHg) | |
Authentic | |
Glass bottle | |
1.6350 to 1.6370 | |
MFCD00004815 | |
09, IV, 777 | |
15, 1117 | |
Solubility in water: almost insoluble. Other solubilities: soluble in chloroform,carbon tetrachloride,,dioxane | |
C1=CC=C(C=C1)C(=O)N=C=S | |
163.2 | |
163.2 | |
Liquid |
97.5 | |
1.2100g/mL | |
>110°C | |
97.5% min. (GC) | |
C8H5NOS | |
C6H5CONCS | |
5 g | |
1.21 | |
benzoylthiocarbimide, benzoylisothiocyanate, benzoylisothiocyante, isothiocyanic acid benzoyl ester, unii-u8j7q32b86, n-benzoyl isothiocyanate, benzoic acid, anhydride with hncs, benzoic acid, anhydride with isothiocyanic acid, oxophenylmethanisothiocyanate, 1-benzoyl acetone | |
CPEKAXYCDKETEN-UHFFFAOYSA-N | |
benzoyl isothiocyanate | |
68284 | |
98% | |
Benzoyl isothiocyanate |
Safety and Handling
GHS H Statement
Causes serious eye damage.
Toxic if swallowed.
May cause an allergic skin reaction.
Contact with acids liberates very toxic gas.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately
GHS Signal Word: Danger
EINECSNumber : 208-540-
RUO – Research Use Only