Please login to your online account to display your discounted pricing

Alfa Aesar™ Benzoyl isocyanate, tech. 90%

Chemical Identifiers

Chemical Name or Material Benzoyl isocyanate
CAS 4461-33-0
Purity Grade Technical Grade
Molecular Formula C8H5NO2
Formula Weight 147.13
MDL Number MFCD00002032
UN Number UN2206
Beilstein 878595
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1158303

  SDS
alfa aesar™
A1158303
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1158306

  SDS
alfa aesar™
A1158306
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 147.13
Molecular Formula C8H5NO2
UN Number UN2206
MDL Number MFCD00002032
Flash Point 72°C (161°F)
Health Hazard 1 Danger
Quantity 1g
CAS 4461-33-0
Purity Grade Technical Grade
Beilstein 878595
Chemical Name or Material Benzoyl isocyanate
Boiling Point 48° to 49°C (0.9mmHg)
Odor Pungent
Health Hazard 2 GHS H Statement
H331-H227-H302-H312-H315-H319-H335
Toxic if inhaled.
Combustible liquid.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Recommended Storage −30°C to −10°C
DOT Information Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: ISOCYANATES, TOXIC, N.O.S.
Health Hazard 3 GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Assay 90%
Safety and Handling

Danger

GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H331-H227-H302-H312-H315-H319-H335
Toxic if inhaled.
Combustible liquid.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

−30°C to −10°C