Learn More
Benzoyl fluoride, 97%, Thermo Scientific Chemicals
$62.64 - $62.64
Chemical Identifiers
CAS | 455-32-3 |
---|---|
Molecular Formula | C7H5FO |
Molecular Weight (g/mol) | 124.11 |
MDL Number | MFCD00000364 |
InChI Key | HPMLGNIUXVXALD-UHFFFAOYSA-N |
Synonym | benzoylfluoride, unii-lor25i34hd, lor25i34hd, benzoylfluorid, acmc-1alcy, hpmlgniuxvxald-uhfffaoysa |
PubChem CID | 67999 |
IUPAC Name | benzoyl fluoride |
SMILES | FC(=O)C1=CC=CC=C1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAL1146406
|
Thermo Scientific Chemicals
L1146406 |
5 g |
Each for $62.64
|
|
|||||
Description
Benzoyl fluoride is used as a mild fluorinating agent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsBenzoyl fluoride is used as a mild fluorinating agent.
Solubility
Hydrolyses in water.
Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers. It is sensitive to moisture. Store at room temperature.
Chemical Identifiers
455-32-3 | |
124.11 | |
HPMLGNIUXVXALD-UHFFFAOYSA-N | |
67999 | |
FC(=O)C1=CC=CC=C1 |
C7H5FO | |
MFCD00000364 | |
benzoylfluoride, unii-lor25i34hd, lor25i34hd, benzoylfluorid, acmc-1alcy, hpmlgniuxvxald-uhfffaoysa | |
benzoyl fluoride |
Specifications
455-32-3 | |
1.14 | |
48°C (118°F) | |
C7H5FO | |
MFCD00000364 | |
UN2920 | |
Moisture sensitive | |
Hydrolyses in water. | |
FC(=O)C1=CC=CC=C1 | |
124.11 | |
124.12 | |
Benzoyl fluoride |
∼-28°C | |
159°C to 161°C | |
Pungent | |
1.496 | |
5 g | |
1904977 | |
benzoylfluoride, unii-lor25i34hd, lor25i34hd, benzoylfluorid, acmc-1alcy, hpmlgniuxvxald-uhfffaoysa | |
HPMLGNIUXVXALD-UHFFFAOYSA-N | |
benzoyl fluoride | |
67999 | |
98% |
Safety and Handling
GHS H Statement
H314-H318-H226
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.
P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
H226-H314-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUIDS, FLAMMABLE, N.O.S.
EINECSNumber : 207-244-9
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only