missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Benzoyl-DL-methionine, TCI America™

$56.53 - $233.75

Chemical Identifiers

CAS 4703-38-2
Molecular Formula C12H15NO3S
Molecular Weight (g/mol) 253.316
MDL Number MFCD00026044
InChI Key PPFRJEXUPZWQPI-UHFFFAOYSA-N
PubChem CID 295480
IUPAC Name 2-benzamido-4-methylsulfanylbutanoic acid
SMILES CSCCC(C(=O)O)NC(=O)C1=CC=CC=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B03001G View Documents TCI America
B03001G
1g Each for $56.53
 
B030010G View Documents TCI America
B030010G
10g Each for $233.75
 
Specifications

Chemical Identifiers

4703-38-2
253.316
PPFRJEXUPZWQPI-UHFFFAOYSA-N
2-benzamido-4-methylsulfanylbutanoic acid
C12H15NO3S
MFCD00026044
295480
CSCCC(C(=O)O)NC(=O)C1=CC=CC=C1

Specifications

White
1g
C12H15NO3S
PPFRJEXUPZWQPI-UHFFFAOYSA-N
2-benzamido-4-methylsulfanylbutanoic acid
295480
Crystal-Powder at 20°C
Benzoyl-DL-methionine
152°C
4703-38-2
MFCD00026044
CSCCC(C(=O)O)NC(=O)C1=CC=CC=C1
253.316
253.32
≥99.0% (T)
Safety and Handling

Safety and Handling

TSCA : No