missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Benzoyl-DL-leucine, TCI America™

$58.06 - $113.06

Chemical Identifiers

CAS 17966-67-5
Molecular Formula C13H17NO3
Molecular Weight (g/mol) 235.283
MDL Number MFCD00038295
InChI Key POLGZPYHEPOBFG-UHFFFAOYSA-N
PubChem CID 294887
IUPAC Name 2-benzamido-4-methylpentanoic acid
SMILES CC(C)CC(C(=O)O)NC(=O)C1=CC=CC=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B020610G View Documents TCI America
B020610G
10g Each for $58.06
 
B020625G View Documents TCI America
B020625G
25g Each for $113.06
 
Specifications

Chemical Identifiers

17966-67-5
235.283
POLGZPYHEPOBFG-UHFFFAOYSA-N
2-benzamido-4-methylpentanoic acid
C13H17NO3
MFCD00038295
294887
CC(C)CC(C(=O)O)NC(=O)C1=CC=CC=C1

Specifications

White-Yellow
10g
C13H17NO3
POLGZPYHEPOBFG-UHFFFAOYSA-N
2-benzamido-4-methylpentanoic acid
294887
Crystal-Powder at 20°C
Benzoyl-DL-leucine
141°C
17966-67-5
MFCD00038295
CC(C)CC(C(=O)O)NC(=O)C1=CC=CC=C1
235.283
235.28
≥99.0% (HPLC,T)
Safety and Handling

Safety and Handling

TSCA : No