Please login to your online account to display your discounted pricing

Benzoyl Cyanide 95%, ACROS Organics™


Quantity 100g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC383661000

 View more versions of this product

Catalog No. AC383661000

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥94%
Physical Form Low Melting Mass
Formula Weight 131.13
Quantity 100g
Color Yellow to brown
Melting Point 27° to 32°C
CAS Min % 94.0
CAS Max % 100.0
Molecular Formula C8H5NO
Linear Formula C6H5COCN
Infrared Spectrum Authentic
Packaging Glass bottle
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Harmful if inhaled.
Toxic in contact with skin.
Toxic if swallowed.
Causes severe skin burns and eye damage.
Very toxic to aquatic life.
Chemical Name or Material Benzoyl cyanide
Assay Percent Range 95%
MDL Number MFCD00001837
CAS 613-90-1
Synonym Phenylglyoxylonitrile
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Immediately call a POISON CENTER or doctor/physician.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material Benzoyl cyanide
CAS 613-90-1
CAS Min % 94.0
CAS Max % 100.0
Assay ≥94%
Assay Percent Range 95%
Molecular Formula C8H5NO
Linear Formula C6H5COCN
Formula Weight 131.13
MDL Number MFCD00001837
Synonym Phenylglyoxylonitrile
Accessories

GHS Signal Word: Danger

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Immediately call a POISON CENTER or doctor/physician.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if inhaled.
Toxic in contact with skin.
Toxic if swallowed.
Causes severe skin burns and eye damage.
Very toxic to aquatic life.