missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Benzoyl Cyanide 95%, ACROS Organics™

$67.58 - $209.97

Chemical Identifiers

CAS 613-90-1
CAS Min % 94.0
CAS Max % 100.0
Assay Percent Range 95%
Molecular Formula C8H5NO
Linear Formula C6H5COCN
Formula Weight 131.13
MDL Number MFCD00001837
Synonym Phenylglyoxylonitrile
Chemical Name or Material Benzoyl cyanide
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC383661000 Thermo Fisher Scientific
AC383661000
Glass bottle 100g Each for $209.97
AC383660250 Thermo Fisher Scientific
AC383660250
Glass bottle 25g Each for $67.58
Specifications Description & Specifications

Chemical Identifiers

613-90-1
100.0
C8H5NO
131.13
Phenylglyoxylonitrile
≥94%
94.0
95%
C6H5COCN
MFCD00001837
Benzoyl cyanide

Specifications

613-90-1
100.0
95%
C6H5COCN
MFCD00001837
Phenylglyoxylonitrile
Yellow to brown
100g
≥94%
94.0
Low Melting Mass
C8H5NO
131.13
Authentic
Glass bottle
27° to 32°C
Benzoyl cyanide

Safety and Handling

GHS H Statement:
Harmful if inhaled.
Toxic in contact with skin.
Toxic if swallowed.
Causes severe skin burns and eye damage.
Very toxic to aquatic life.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Immediately call a POISON CENTER or doctor/physician.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger