Please login to your online account to display your discounted pricing

Alfa Aesar™ Benzoyl chloride on polystyrene, 0.8-1.0 mmol/g

Chemical Identifiers

Chemical Name or Material Benzoyl chloride on polystyrene
MDL Number MFCD01865658
Synonym 4-Chlorocarbonylpolystyrene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1946003

 
alfa aesar™
L1946003
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Benzoyl chloride on polystyrene
MDL Number MFCD01865658
Concentration 0.8 to 1.0mmol/g
Quantity 1g
Synonym 4-Chlorocarbonylpolystyrene