Learn More
Benzoyl chloride, 99+%, Thermo Scientific Chemicals
$46.49 - $210.47
Chemical Identifiers
CAS | 98-88-4 |
---|---|
Molecular Formula | C7H5ClO |
MDL Number | MFCD00000653 |
InChI Key | PASDCCFISLVPSO-UHFFFAOYSA-N |
Synonym | benzenecarbonyl chloride, benzoic acid, chloride, benzoylchloride, alpha-chlorobenzaldehyde, benzoic acid chloride, benzoylchlorid, benzaldehyde, alpha-chloro, a-chlorobenzaldehyde, ccris 802, unii-vty8706w36 |
PubChem CID | 7412 |
ChEBI | CHEBI:82275 |
IUPAC Name | benzoyl chloride |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1410730
|
Thermo Scientific Chemicals
A1410730 |
250 g |
Each for $46.49
|
|
|||||
AAA141070B
|
Thermo Scientific Chemicals
A141070B |
1000 g |
Each for $87.66
|
|
|||||
AAA141070E
|
Thermo Scientific Chemicals
A141070E |
2500 g |
Each for $210.47
|
|
|||||
Description
Benzoyl chloride is widely utilized for the synthesis of peroxides. It is employed in the production of dyes and perfumes. It also serves in the manufacturing of pharmaceuticals and resins.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsBenzoyl chloride is widely utilized for the synthesis of peroxides. It is employed in the production of dyes and perfumes. It also serves in the manufacturing of pharmaceuticals and resins.
Solubility
Miscible with water, ether, benzene, carbon disulfide and carbon tetra chloride.
Notes
Moisture sensitive. Incompatible with strong oxidizing agents, strong bases and alcohols.
Chemical Identifiers
98-88-4 | |
MFCD00000653 | |
benzenecarbonyl chloride, benzoic acid, chloride, benzoylchloride, alpha-chlorobenzaldehyde, benzoic acid chloride, benzoylchlorid, benzaldehyde, alpha-chloro, a-chlorobenzaldehyde, ccris 802, unii-vty8706w36 | |
CHEBI:82275 |
C7H5ClO | |
PASDCCFISLVPSO-UHFFFAOYSA-N | |
7412 | |
benzoyl chloride |
Specifications
98-88-4 | |
1.211 | |
69°C (156°F) | |
C7H5ClO | |
MFCD00000653 | |
UN1736 | |
14,1112 | |
Miscible with water,ether,benzene,carbon disulfide and carbon tetra chloride. | |
C1=CC=C(C=C1)C(=O)Cl | |
140.566 | |
CHEBI:82275 | |
≥99% | |
Benzoyl Chloride |
°C to 1°C | |
197°C to 199°C | |
≥99% | |
1.5535 | |
250 g | |
Moisture sensitive | |
benzenecarbonyl chloride, benzoic acid, chloride, benzoylchloride, alpha-chlorobenzaldehyde, benzoic acid chloride, benzoylchlorid, benzaldehyde, alpha-chloro, a-chlorobenzaldehyde, ccris 802, unii-vty8706w36 | |
PASDCCFISLVPSO-UHFFFAOYSA-N | |
benzoyl chloride | |
7412 | |
140.57 | |
Liquid |
Safety and Handling
GHS H Statement
H314-H302-H312-H332-H317-H227
P210-P235-P260-P264b-P270-P271-P272-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P333+P313-P363-P370+P378q-P501c
H227-H302-H311+H331-H314-H317-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: BENZOYL CHLORIDE
EINECSNumber : 202-710-8
TSCA : Yes
storageNote1 : Moisture sensitive
Recommended Storage : Ambient temperatures
RUO – Research Use Only