missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Benzoyl chloride, 99+%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A141070B

 View more versions of this product

Catalog No. AAA141070B



Description

Description

Benzoyl chloride is widely utilized for the synthesis of peroxides. It is employed in the production of dyes and perfumes. It also serves in the manufacturing of pharmaceuticals and resins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Benzoyl chloride is widely utilized for the synthesis of peroxides. It is employed in the production of dyes and perfumes. It also serves in the manufacturing of pharmaceuticals and resins.

Solubility
Miscible with water, ether, benzene, carbon disulfide and carbon tetra chloride.

Notes
Moisture sensitive. Incompatible with strong oxidizing agents, strong bases and alcohols.
Specifications

Specifications

Benzoyl chloride
-1°C
2
69°C (156°F)
Pungent
1.5535
1000 g
Moisture sensitive
benzenecarbonyl chloride, benzoic acid, chloride, benzoylchloride, alpha-chlorobenzaldehyde, benzoic acid chloride, benzoylchlorid, benzaldehyde, alpha-chloro, a-chlorobenzaldehyde, ccris 802, unii-vty8706w36
PASDCCFISLVPSO-UHFFFAOYSA-N
benzoyl chloride
7412
140.57
Liquid
98-88-4
1.211
197°C to 199°C
≥99%
C7H5ClO
MFCD00000653
UN1736
14,1112
Miscible with water,ether,benzene,carbon disulfide and carbon tetra chloride.
C1=CC=C(C=C1)C(=O)Cl
140.566
CHEBI:82275
≥99%

Chemical Identifiers

98-88-4
MFCD00000653
benzenecarbonyl chloride, benzoic acid, chloride, benzoylchloride, alpha-chlorobenzaldehyde, benzoic acid chloride, benzoylchlorid, benzaldehyde, alpha-chloro, a-chlorobenzaldehyde, ccris 802, unii-vty8706w36
CHEBI:82275
C7H5ClO
PASDCCFISLVPSO-UHFFFAOYSA-N
7412
benzoyl chloride
Safety and Handling

Safety and Handling

GHS H Statement
H314-H302-H312-H332-H317-H227
Causes severe skin burns and eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
May cause an allergic skin reaction.
Combustible liquid.

P210-P235-P260-P264b-P270-P271-P272-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P333+P313-P363-P370+P378q-P501c

H227-H302-H311+H331-H314-H317-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: BENZOYL CHLORIDE

EINECSNumber : 202-710-8

TSCA : Yes

storageNote1 : Moisture sensitive

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only