missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Benzoyl bromide, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC160950250

 View more versions of this product

Catalog No. AC160950250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Benzoyl bromide
90°C
Glass bottle
1.57
Orange
25mL
96.0
96% min. (GC)
C6H5COBr
09,195
Solubility in water: may decompose. Other solubilities: miscible in ethyl ether
C1=CC=C(C=C1)C(=O)Br
185.02
185.02
97%
1.5700g/mL
Authentic
1.5880 to 1.5900
218.0°C to 219.0°C
-24.0°C
618-32-6
100.0
C7H5BrO
MFCD00000112
6ci,7ci,8ci,9ci
AQIHMSVIAGNIDM-UHFFFAOYSA-N
benzoyl bromide
12056
Liquid

Chemical Identifiers

618-32-6
185.02
AQIHMSVIAGNIDM-UHFFFAOYSA-N
12056
C1=CC=C(C=C1)C(=O)Br
C7H5BrO
MFCD00000112
6ci,7ci,8ci,9ci
benzoyl bromide
Safety and Handling

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Reacts violently with water.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 210-544-2

SDS Documents