Please login to your online account to display your discounted pricing

Alfa Aesar™ Benzoyl(2-hydroxybenzoyl)methane, 98%

$29.99 - $144.80

Chemical Identifiers

Chemical Name or Material Benzoyl(2-hydroxybenzoyl)methane
CAS 1469-94-9
Molecular Formula C15H12O3
Formula Weight 240.26
MDL Number MFCD00043495
Synonym 1-(2-Hydroxyphenyl)-3-phenyl-1,3-propanedione
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2917603

  SDS
alfa aesar™
H2917603
1g Each for $29.99

AAH2917609

  SDS
alfa aesar™
H2917609
10g Each for $144.80
Description & Specifications

Specifications

Formula Weight 240.26
Chemical Name or Material Benzoyl(2-hydroxybenzoyl)methane
TSCA TSCA
Assay 98%
Molecular Formula C15H12O3
Melting Point 118° to 122°C
MDL Number MFCD00043495
Quantity 1g
CAS 1469-94-9
Synonym 1-(2-Hydroxyphenyl)-3-phenyl-1,3-propanedione
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.