missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Benzoxazole, 97+%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 105680250

 View more versions of this product

Catalog No. AC105680250



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Benzoxazole
273-53-0
100.0
Brown-Orange to Yellow
58°C
97% min. (GC)
C7H5NO
25 g
benzoxazole, 1-oxa-3-azaindene, 1-oxa-3-aza-1h-indene, benzo d oxazole, benzooxazole, usaf ek-5017, unii-j233y1i55i, benzoxazol, benzoxazoline, benzothiazoie
BCMCBBGGLRIHSE-UHFFFAOYSA-N
1,3-benzoxazole
9228
119.12
Liquid
99+%
99.0
27°C to 30°C
182°C
Authentic
Glass bottle
MFCD00005765
27, 42
Solubility in water: insoluble. Other solubilities: soluble in ethanol,sulfuric acid
O1C=NC2=CC=CC=C12
119.12
CHEBI:38814
97+%

Chemical Identifiers

273-53-0
119.12
BCMCBBGGLRIHSE-UHFFFAOYSA-N
9228
1,3-benzoxazole
C7H5NO
MFCD00005765
benzoxazole, 1-oxa-3-azaindene, 1-oxa-3-aza-1h-indene, benzo d oxazole, benzooxazole, usaf ek-5017, unii-j233y1i55i, benzoxazol, benzoxazoline, benzothiazoie
CHEBI:38814
O1C=NC2=CC=CC=C12
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
Harmful if swallowed.
May cause respiratory irritation.
Flammable solid.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep container tightly closed.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 205-988-9

RTECSNumber : DM4375000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only