missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Benzotrichloride, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A156520E

 View more versions of this product

Catalog No. AAA156520E



Description

Description

It finds its use in other preparations of substituted benzophenones, for benzoylation of phenols under basic, phase-transfer conditions. D,D,D-Trichlorotoluene is an intermediate, used exclusively in the industrial production of other intermediates such as Benzoylchloride, Benzotrifluoride and 2,4-Dihydroxybenzophenone. α,α,α-Trichlorotoluene is also used in determination of chlorinated toluenes in effluents of wastewater treatment plants and textile industries by closed loop stripping analysis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It finds its use in other preparations of substituted benzophenones, for benzoylation of phenols under basic, phase-transfer conditions. D,D,D-Trichlorotoluene is an intermediate, used exclusively in the industrial production of other intermediates such as Benzoylchloride, Benzotrifluoride and 2,4-Dihydroxybenzophenone. α,α,α-Trichlorotoluene is also used in determination of chlorinated toluenes in effluents of wastewater treatment plants and textile industries by closed loop stripping analysis.

Solubility
Solubility in water is 0.1 g/L at 20°C.

Notes
Special handling precautions required. View MSDS prior to purchase. MSDS are available online at www.alfa.com
Specifications

Specifications

Benzotrichloride
-8°C to -0.5°C
90°C to 91°C (15 mmHg)
C7H5Cl3
MFCD00000786
UN2226
Moisture sensitive
benzotrichloride, trichloromethyl benzene, alpha,alpha,alpha-trichlorotoluene, phenylchloroform, toluene trichloride, benzenyl trichloride, benzene, trichloromethyl, benzyl trichloride, benzylidyne chloride, phenyltrichloromethane
ClC(Cl)(Cl)C1=CC=CC=C1
195.47
CHEBI:82274
98%
98-07-7
1.38
97°C (207°F)
1.557
2500 g
508152
14,1109
XEMRAKSQROQPBR-UHFFFAOYSA-N
trichloromethylbenzene
7367
195.48

Chemical Identifiers

98-07-7
195.47
XEMRAKSQROQPBR-UHFFFAOYSA-N
7367
trichloromethylbenzene
C7H5Cl3
MFCD00000786
benzotrichloride, trichloromethyl benzene, alpha,alpha,alpha-trichlorotoluene, phenylchloroform, toluene trichloride, benzenyl trichloride, benzene, trichloromethyl, benzyl trichloride, benzylidyne chloride, phenyltrichloromethane
CHEBI:82274
ClC(Cl)(Cl)C1=CC=CC=C1
Safety and Handling

Safety and Handling

GHS H Statement
H330-H350-H314-H318-H302-H335
Fatal if inhaled.
May cause cancer.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
May cause respiratory irritation.

P201-P202-P261-P264b-P270-P271-P281-P301+P312-P302+P352-P304+P340-P305+P351+P338-P308+P313-P310-P311-P330-P332+P313-P362-P501c

H302-H315-H318-H331-H335-H350

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: BENZOTRICHLORIDE

EINECSNumber : 202-634-5

RTECSNumber : XT9275000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only