missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Benzotriazol-1-yloxytris(dimethylamino)-phosphonium hexafluorophosphate, 98+%, ACROS Organics™

Manufacturer:  Acros Organics 209800050

 View more versions of this product

Catalog No. AC209800050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Coupling reagent
Specifications

Specifications

Benzotriazol-1-yloxytris(dimethylamino)-phosphonium hexafluorophosphate
Glass bottle
136.0°C to 140.0°C
56602-33-6
C12H22F6N6OP2
bop reagent, 1h-benzo d 1,2,3 triazol-1-yl oxy tris dimethylamino phosphonium hexafluorophosphate v, castro's reagent, bop-reagent, benzotriazol-1-yloxytris dimethylamino-phosphonium hexafluorophosphate, ccris 2602, castros reagent, tri dimethylamino benzotriazol-1-yloxyphosphonium hexafluorophosphate, benzotriazol-1-yloxy tris dimethylamino phosphonium hexafluorophosphate, 1h-benzotriazol-1-yloxytris dimethylamino phosphonium hexafluorophosphate
CN(C)[P+](N(C)C)(N(C)C)ON1C2=CC=CC=C2N=N1.F[P-](F)(F)(F)(F)F
442.287
442.27
≥98%
Authentic
White
5g
98+%
06,34; 07,14; 11,68
MGEVGECQZUIPSV-UHFFFAOYSA-N
benzotriazol-1-yloxy-tris(dimethylamino)phosphanium;hexafluorophosphate
151348
Powder

Chemical Identifiers

56602-33-6
442.287
bop reagent, 1h-benzo d 1,2,3 triazol-1-yl oxy tris dimethylamino phosphonium hexafluorophosphate v, castro's reagent, bop-reagent, benzotriazol-1-yloxytris dimethylamino-phosphonium hexafluorophosphate, ccris 2602, castros reagent, tri dimethylamino benzotriazol-1-yloxyphosphonium hexafluorophosphate, benzotriazol-1-yloxy tris dimethylamino phosphonium hexafluorophosphate, 1h-benzotriazol-1-yloxytris dimethylamino phosphonium hexafluorophosphate
benzotriazol-1-yloxy-tris(dimethylamino)phosphanium;hexafluorophosphate
C12H22F6N6OP2
MGEVGECQZUIPSV-UHFFFAOYSA-N
151348
CN(C)[P+](N(C)C)(N(C)C)ON1C2=CC=CC=C2N=N1.F[P-](F)(F)(F)(F)F
SDS Documents