missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Benzotriazol-1-yloxytris(dimethylamino)-phosphonium hexafluorophosphate, 98+%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 209800010

 View more versions of this product

Catalog No. AC209800010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Coupling reagent
Specifications

Specifications

Benzotriazol-1-yloxytris(dimethylamino)-phosphonium hexafluorophosphate
Glass bottle
136.0°C to 140.0°C
56602-33-6
C12H22N6OP·PF6
1h-benzo d 1,2,3 triazol-1-yl oxy tris dimethylamino phosphonium hexafluorophosphate v, 1h-benzotriazol-1-yloxytris dimethylamino phosphonium hexafluorophosphate, benzotriazol-1-yloxy tris dimethylamino phosphonium hexafluorophosphate, benzotriazol-1-yloxytris dimethylamino-phosphonium hexafluorophosphate, bop reagent, bop-reagent, castro's reagent, castros reagent, ccris 2602, tri dimethylamino benzotriazol-1-yloxyphosphonium hexafluorophosphate
CN(C)[P+](N(C)C)(N(C)C)ON1C2=CC=CC=C2N=N1.F[P-](F)(F)(F)(F)F
442.27
442.27
98+%
Authentic
White
1g
98% min. (HPLC)
06,34; 07,14
MGEVGECQZUIPSV-UHFFFAOYSA-N
benzotriazol-1-yloxy-tris(dimethylamino)phosphanium;hexafluorophosphate
151348
Powder

Chemical Identifiers

56602-33-6
442.27
1h-benzo d 1,2,3 triazol-1-yl oxy tris dimethylamino phosphonium hexafluorophosphate v, 1h-benzotriazol-1-yloxytris dimethylamino phosphonium hexafluorophosphate, benzotriazol-1-yloxy tris dimethylamino phosphonium hexafluorophosphate, benzotriazol-1-yloxytris dimethylamino-phosphonium hexafluorophosphate, bop reagent, bop-reagent, castro's reagent, castros reagent, ccris 2602, tri dimethylamino benzotriazol-1-yloxyphosphonium hexafluorophosphate
benzotriazol-1-yloxy-tris(dimethylamino)phosphanium;hexafluorophosphate
C12H22N6OP·PF6
MGEVGECQZUIPSV-UHFFFAOYSA-N
151348
CN(C)[P+](N(C)C)(N(C)C)ON1C2=CC=CC=C2N=N1.F[P-](F)(F)(F)(F)F
Safety and Handling

Safety and Handling

GHS H Statement
Causes serious eye irritation.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attention.
IF INHALED: Remove to fresh air and keep

GHS Signal Word: Danger

EINECSNumber : 260-279-1

SDS Documents