missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(Benzotriazol-1-yloxy)dipiperidinocarbenium hexafluorophosphate, 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC440240010

 View more versions of this product

Catalog No. AC440240010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

(Benzotriazol-1-yloxy)dipiperidinocarbenium hexafluorophosphate
Authentic
183°C to 190°C
190849-64-0
100.0
C17H24N5O·F6P
YNOBMGHLCWIWCL-UHFFFAOYSA-N
1-[piperidin-1-ium-1-ylidene(piperidin-1-yl)methoxy]benzotriazole;hexafluorophosphate
10863473
Powder
43.34 to 45.56% C, 14.86 to 15.63% N
White
1g
97.5
98%
1,2,3-benzotriazol-1-yl bis piperidin-1-yl methylidene oxidanium hexafluorophosphate, 1,2,3-benzotriazol-1-yloxy bis piperidin-1-yl methylium hexafluorophosphate, 1-1h-benzo d 1,2,3 triazol-1-yl oxy piperidin-1-yl methylene piperidin-1-ium hexafluorophosphate v, 1-bis 1-piperidyl methoxy benzotriazole hexafluorophosphate, 1-piperidin-1-ium-1-ylidene 1-piperidyl methoxy benzotriazole hexafluorophosphate, benzotriazol-1-yloxy dipiperidinocarbenium hexafluorophosphate, hbpipu, hbpipu, benzotriazol-1-yloxy dipiperidinocarbenium hexafluorophosphate, o-benzotriazol-1-yl-n,n,n',n'-bis pentamethylene uronium hexafluorophosphate, o-benzotriazol-1-yl-n,n,n',n'-bis pentamethylene uroniumhexafluorophosphate
C1CCN(CC1)C(=[N+]2CCCCC2)ON3C4=CC=CC=C4N=N3.F[P-](F)(F)(F)(F)F
459.38
459.38
98%

Chemical Identifiers

190849-64-0
459.38
1,2,3-benzotriazol-1-yl bis piperidin-1-yl methylidene oxidanium hexafluorophosphate, 1,2,3-benzotriazol-1-yloxy bis piperidin-1-yl methylium hexafluorophosphate, 1-1h-benzo d 1,2,3 triazol-1-yl oxy piperidin-1-yl methylene piperidin-1-ium hexafluorophosphate v, 1-bis 1-piperidyl methoxy benzotriazole hexafluorophosphate, 1-piperidin-1-ium-1-ylidene 1-piperidyl methoxy benzotriazole hexafluorophosphate, benzotriazol-1-yloxy dipiperidinocarbenium hexafluorophosphate, hbpipu, hbpipu, benzotriazol-1-yloxy dipiperidinocarbenium hexafluorophosphate, o-benzotriazol-1-yl-n,n,n',n'-bis pentamethylene uronium hexafluorophosphate, o-benzotriazol-1-yl-n,n,n',n'-bis pentamethylene uroniumhexafluorophosphate
1-[piperidin-1-ium-1-ylidene(piperidin-1-yl)methoxy]benzotriazole;hexafluorophosphate
C17H24N5O·F6P
YNOBMGHLCWIWCL-UHFFFAOYSA-N
10863473
C1CCN(CC1)C(=[N+]2CCCCC2)ON3C4=CC=CC=C4N=N3.F[P-](F)(F)(F)(F)F
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS Documents