missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(Benzotriazol-1-yloxy)dipiperidinocarbenium hexafluorophosphate, 98%, ACROS Organics™

Chemical Identifiers

CAS 190849-64-0
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Molecular Formula C17H24N5O·F6P
Formula Weight 459.38
Synonym HBPipU
Chemical Name or Material (Benzotriazol-1-yloxy)dipiperidinocarbenium hexafluorophosphate
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC440240010 SDS Thermo Fisher Scientific
AC440240010
1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC440240050 SDS Thermo Fisher Scientific
AC440240050
5g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

190849-64-0
100.0
C17H24N5O·F6P
HBPipU
97.5
98%
459.38
(Benzotriazol-1-yloxy)dipiperidinocarbenium hexafluorophosphate

Specifications

Powder
1g
100.0
97.5
C17H24N5O·F6P
43.34 to 45.56% C, 14.86 to 15.63% N
(Benzotriazol-1-yloxy)dipiperidinocarbenium hexafluorophosphate
459.38
White to off-white
Authentic
98%
190849-64-0
HBPipU

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.