Please login to your online account to display your discounted pricing

Benzoquinonium dibromide, Tocris


Nicotinic antagonist

Quantity 10mg

Manufacturer: r & d systems  RDS042410

 View more versions of this product

Catalog No. 042410

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 10mg
Formula Weight 706.6
CAS 311-09-1
Recommended Storage Store at Room Temperature
Synonym N,N'-[(3,6-Dioxo-1,4-cyclohexadiene-1,4-diyl)bis(imino-3,1-propanediyl)]bis[N,N-diethylbenzenemethanaminium]dibromide
Molecular Formula C34H50Br2N4O2
Chemical Name or Material Benzoquinonium dibromide
Assay Percent Range >98%

Specifications

Chemical Name or Material Benzoquinonium dibromide
Assay Percent Range >98%
Molecular Formula C34H50Br2N4O2
Formula Weight 706.6
Synonym N,N'-[(3,6-Dioxo-1,4-cyclohexadiene-1,4-diyl)bis(imino-3,1-propanediyl)]bis[N,N-diethylbenzenemethanaminium]dibromide
Accessories

Store at Room Temperature