Please login to your online account to display your discounted pricing

Benzoquinonium dibromide, Tocris

Nicotinic antagonist

$164.61 - $682.67

Chemical Identifiers

Chemical Name or Material Benzoquinonium dibromide
Assay Percent Range >98%
Molecular Formula C34H50Br2N4O2
Formula Weight 706.6
Synonym N,N'-[(3,6-Dioxo-1,4-cyclohexadiene-1,4-diyl)bis(imino-3,1-propanediyl)]bis[N,N-diethylbenzenemethanaminium]dibromide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

042410

 
r & d systems
RDS042410
10mg Each for $164.61

042450

 
r & d systems
RDS042450
50mg Each for $682.67
Description & Specifications

Specifications

Quantity 10mg
Formula Weight 706.6
CAS 311-09-1
Recommended Storage Store at Room Temperature
Chemical Name or Material Benzoquinonium dibromide
Assay Percent Range >98%
Molecular Formula C34H50Br2N4O2
Synonym N,N'-[(3,6-Dioxo-1,4-cyclohexadiene-1,4-diyl)bis(imino-3,1-propanediyl)]bis[N,N-diethylbenzenemethanaminium]dibromide
Safety and Handling

Store at Room Temperature