missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Benzopinacolone, 97%

$42.72 - $178.89

Chemical Identifiers

CAS 466-37-5
Molecular Formula C26H20O
Molecular Weight (g/mol) 348.445
MDL Number MFCD00004762
InChI Key CFBBKHROQRFCNZ-UHFFFAOYSA-N
Synonym 2,2,2-triphenylacetophenone, tetraphenylethanone, benzopinacolone, ethanone, tetraphenyl, phenyl trityl ketone, acetophenone, 2,2,2-triphenyl, 1,2,2,2-tetraphenylethan-1-one, ss-benzpinakolin, beta-benzopinacolone, .beta.-benzopinacolone
PubChem CID 68049
IUPAC Name 1,2,2,2-tetraphenylethanone
SMILES C1=CC=C(C=C1)C(=O)C(C2=CC=CC=C2)(C3=CC=CC=C3)C4=CC=CC=C4
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL0169506 View Documents Alfa Aesar™
L0169506
5g Each for $42.72
 
AAL0169514 View Documents Alfa Aesar™
L0169514
25g Each for $178.89
 
Specifications

Chemical Identifiers

466-37-5
348.445
CFBBKHROQRFCNZ-UHFFFAOYSA-N
68049
C1=CC=C(C=C1)C(=O)C(C2=CC=CC=C2)(C3=CC=CC=C3)C4=CC=CC=C4
C26H20O
MFCD00004762
2,2,2-triphenylacetophenone, tetraphenylethanone, benzopinacolone, ethanone, tetraphenyl, phenyl trityl ketone, acetophenone, 2,2,2-triphenyl, 1,2,2,2-tetraphenylethan-1-one, ss-benzpinakolin, beta-benzopinacolone, .beta.-benzopinacolone
1,2,2,2-tetraphenylethanone

Specifications

181°C to 182°C
466-37-5
MFCD00004762
2,2,2-triphenylacetophenone, tetraphenylethanone, benzopinacolone, ethanone, tetraphenyl, phenyl trityl ketone, acetophenone, 2,2,2-triphenyl, 1,2,2,2-tetraphenylethan-1-one, ss-benzpinakolin, beta-benzopinacolone, .beta.-benzopinacolone
C1=CC=C(C=C1)C(=O)C(C2=CC=CC=C2)(C3=CC=CC=C3)C4=CC=CC=C4
348.445
348.45
Benzopinacolone
5g
C26H20O
2058376
CFBBKHROQRFCNZ-UHFFFAOYSA-N
1,2,2,2-tetraphenylethanone
68049
97%