Please login to your online account to display your discounted pricing

Alfa Aesar™ Benzopinacol, 98%

$74.28 - $74.28

Chemical Identifiers

Chemical Name or Material Benzopinacol
CAS 464-72-2
Molecular Formula C26H22O2
Formula Weight 366.46
MDL Number MFCD00004448
Beilstein 2061947
Merck Index 14,1100
Synonym 1,1,2,2-Tetraphenyl-1,2-ethanediol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1142114

  SDS
alfa aesar™
A1142114
25g Each for $74.28
Description & Specifications

Specifications

Formula Weight 366.46
Chemical Name or Material Benzopinacol
TSCA TSCA
Assay 98%
Molecular Formula C26H22O2
MDL Number MFCD00004448
CAS 464-72-2
Beilstein 2061947
Merck Index 14,1100
Synonym 1,1,2,2-Tetraphenyl-1,2-ethanediol
Melting Point 174° to 180°C
Quantity 25g
RTECS Number KI2750000

Applications
Benzopinacol is a catalyst of the formation of unsaturated polyesters. It is also used as an initiator of polymerisation by free radicals. It is also used as an organic synthesis intermediate.

Solubility
It is soluble in ethanol, ether, acetone insoluble in carbon disulfide and water.

Notes
Keep container tightly closed in a dry and well-ventilated place. Store in cool place. Stable under recommended storage conditions. Incompatible to Strong oxidizing agents.