missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Benzophenone p-toluenesulfonylhydrazone, 97%

Manufacturer:  Alfa Aesar™ H5348622

 View more versions of this product

Catalog No. AAH5348622



Specifications

Specifications

Benzophenone p-toluenesulfonylhydrazone
100g
C20H18N2O2S
benzophenone p-toluenesulfonylhydrazone
CC1=CC=C(C=C1)S(=O)(=O)NN=C(C2=CC=CC=C2)C3=CC=CC=C3
350.436
350.44
178°C to 182°C
4545-20-4
MFCD00381345
AVWHDUALELPLAI-UHFFFAOYSA-N
N-(benzhydrylideneamino)-4-methylbenzenesulfonamide
274531
97%

Chemical Identifiers

4545-20-4
350.436
AVWHDUALELPLAI-UHFFFAOYSA-N
274531
CC1=CC=C(C=C1)S(=O)(=O)NN=C(C2=CC=CC=C2)C3=CC=CC=C3
C20H18N2O2S
MFCD00381345
benzophenone p-toluenesulfonylhydrazone
N-(benzhydrylideneamino)-4-methylbenzenesulfonamide
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures

Documents