Please login to your online account to display your discounted pricing

Alfa Aesar™ Benzophenone oxime, 97%

$213.96 - $836.26

Chemical Identifiers

Chemical Name or Material Benzophenone oxime
CAS 574-66-3
Molecular Formula C13H11NO
Formula Weight 197.24
MDL Number MFCD00051461
Beilstein 1869643
Merck Index 14,1098
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1403718

 
alfa aesar™
A1403718
50g Each for $213.96

AAA1403730

 
alfa aesar™
A1403730
250g Each for $836.26

AAA1403709

 
alfa aesar™
A1403709
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Benzophenone oxime
CAS 574-66-3
Recommended Storage −30° to −10°C
RTECS Number DJ1810000
TSCA TSCA
Assay 97%
Molecular Formula C13H11NO
Formula Weight 197.24
MDL Number MFCD00051461
Beilstein 1869643
Merck Index 14,1098
Melting Point 138° to 143°C
Quantity 50g
Safety and Handling

−30° to −10°C