missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Benzonitrile, 99+%, for spectroscopy, ACROS Organics™

Manufacturer:  Acros Organics 167672500

 View more versions of this product

Catalog No. AC167672500



Description

Description

For UV/VIS spectroscopy
Specifications

Specifications

(1 cm cell vs water), (1 cm cell vs water) at 0nm, 0.01 max. at 360nm, 0.01 max. at 400nm, 0.03 max. at 335nm, 0.15 max. at 310nm, 0.3 max. at 305nm, 1 max. at 300nm
1.0100g/mL
Glass bottle
0.0005% max.
0.03% max.
Undesignated
250mL
100-47-0
100.0
C7H5N
MFCD00001770
15, 1099
JFDZBHWFFUWGJE-UHFFFAOYSA-N
benzonitrile
7505
103.12
≥99%
Benzonitrile
71°C
1.5270 to 1.5290
1.01
188.0°C to 191.0°C
-13.0°C
99+%
99.0
99+%
C6H5CN
09, 275
phenyl cyanide, cyanobenzene, benzenenitrile, benzoic acid nitrile, benzene, cyano, benzenecarbonitrile, phenylcyanide, fenylkyanid, fenylkyanid czech, benzonitril
C1=CC=C(C=C1)C#N
103.124
CHEBI:27991
Liquid
Spectroscopy

Chemical Identifiers

100-47-0
103.124
JFDZBHWFFUWGJE-UHFFFAOYSA-N
7505
benzonitrile
C7H5N
MFCD00001770
phenyl cyanide, cyanobenzene, benzenenitrile, benzoic acid nitrile, benzene, cyano, benzenecarbonitrile, phenylcyanide, fenylkyanid, fenylkyanid czech, benzonitril
CHEBI:27991
C1=CC=C(C=C1)C#N
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful in contact with skin.
Harmful if swallowed.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning

EINECSNumber : 202-855-7

RTECSNumber : DI2450000

TSCA : TSCA

SDS Documents