missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Benzoin isopropyl ether, 98+%

$58.95 - $724.03

Chemical Identifiers

CAS 6652-28-4
Molecular Formula C17H18O2
Formula Weight 254.33
MDL Number MFCD00015025
Beilstein 1977110
Synonym alpha-Isopropoxy-alpha-phenylacetophenone
Chemical Name or Material Benzoin isopropyl ether
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2265414 SDS Alfa Aesar™
B2265414
25g Each for $58.95
AAB2265422 SDS Alfa Aesar™
B2265422
100g Each for $179.51
AAB2265436 SDS Alfa Aesar™
B2265436
500g Each for $724.03
Specifications Description & Specifications

Chemical Identifiers

6652-28-4
254.33
1977110
Benzoin isopropyl ether
C17H18O2
MFCD00015025
alpha-Isopropoxy-alpha-phenylacetophenone

Specifications

6652-28-4
254.33
1977110
180°C (10mmHg)
25g
≥98%
C17H18O2
MFCD00015025
alpha-Isopropoxy-alpha-phenylacetophenone
76° to 79°C
Benzoin isopropyl ether

Safety and Handling

TSCA : TSCA