Please login to your online account to display your discounted pricing

Alfa Aesar™ Benzoin ethyl ether, 98%

Chemical Identifiers

Chemical Name or Material Benzoin ethyl ether
CAS 574-09-4
Molecular Formula C16H16O2
Formula Weight 240.3
MDL Number MFCD00009242
UN Number UN3077
Beilstein 2053926
Merck Index 14,1093
Synonym 2-Ethoxy-2-phenylacetophenone; O-Ethylbenzoin
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA187820B

  SDS
alfa aesar™
A187820B
1000g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1878218

  SDS
alfa aesar™
A1878218
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1878230

  SDS
alfa aesar™
A1878230
250g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 240.3
Chemical Name or Material Benzoin ethyl ether
TSCA TSCA
Assay 98%
Molecular Formula C16H16O2
MDL Number MFCD00009242
CAS 574-09-4
UN Number UN3077
Beilstein 2053926
Merck Index 14,1093
Synonym 2-Ethoxy-2-phenylacetophenone; O-Ethylbenzoin
Boiling Point 194° to 195°C (20mmHg)
Melting Point 55° to 59°C
Health Hazard 1 Danger
Quantity 1000g
Health Hazard 2 GHS H Statement
H301
Toxic if swallowed.
Health Hazard 3 GHS P Statement
P264-P270-P301+P310-P321-P405-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
Safety and Handling

Danger

GHS P Statement
P264-P270-P301+P310-P321-P405-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301
Toxic if swallowed.