Please login to your online account to display your discounted pricing

Benzoin 98%, ACROS Organics™


Quantity 500g
Packaging Plastic bottle

Manufacturer: fisher scientific acros  105515000

 View more versions of this product

Catalog No. AC105515000

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Physical Form Powder
Formula Weight 212.25
Quantity 500g
Color Off-white to very slightly yellow
Merck Index 15, 1095
RTECS Number DI1590000
TSCA TSCA
EINECS Number 204-331-3
Melting Point 133° to 137°C
CAS Max % 100.0
Sulfated Ash 0.2% max.
Loss on Drying 1% max.
Assay Percent Range 98%
Linear Formula C6H5CH(OH)COC6H5
Molecular Formula C14H12O2
CAS Min % 97.5
Chemical Name or Material Benzoin
Name Note 98%
MDL Number MFCD00004496
Packaging Plastic bottle
CAS 119-53-9
Synonym α-Hydroxy-α-phenylacetophenone
Infrared Spectrum Authentic

Specifications

Chemical Name or Material Benzoin
Name Note 98%
CAS 119-53-9
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C14H12O2
Linear Formula C6H5CH(OH)COC6H5
Formula Weight 212.25
MDL Number MFCD00004496
Synonym α-Hydroxy-α-phenylacetophenone