Please login to your online account to display your discounted pricing

Benzoic Acid USP Crystal, Macron Fine Chemicals™

macron

Chemical Identifiers

Chemical Name or Material Benzoic Acid
CAS 65-85-0
Purity Grade USP
Physical Form Crystalline
Molecular Formula C6H5COOH
Formula Weight 122.12g/mol
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

MK01082

 
avantor performance
010808
Poly 2kg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Packaging Poly
Quantity 2kg
Chemical Name or Material Benzoic Acid
CAS 65-85-0
Purity Grade USP
Physical Form Crystalline
Molecular Formula C6H5COOH
Formula Weight 122.12g/mol