Please login to your online account to display your discounted pricing

Benzoic Acid USP Crystal, Macron Fine Chemicals™


macron

Quantity 2kg
Packaging Poly

Manufacturer: avantor performance  010808

 View more versions of this product

Catalog No. MK01082

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Packaging Poly
Quantity 2kg
Chemical Name or Material Benzoic Acid
CAS 65-85-0
Purity Grade USP
Physical Form Crystalline
Molecular Formula C6H5COOH
Formula Weight 122.12g/mol

Specifications

Chemical Name or Material Benzoic Acid
CAS 65-85-0
Purity Grade USP
Physical Form Crystalline
Molecular Formula C6H5COOH
Formula Weight 122.12g/mol