Please login to your online account to display your discounted pricing

Alfa Aesar™ Benzoic acid on polystyrene, 1.6-2.1 mmol/g

$84.82 - $551.47

Chemical Identifiers

Chemical Name or Material Benzoic acid on polystyrene
MDL Number MFCD03456189
Synonym 4-Carboxypolystyrene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1947303

  SDS
alfa aesar™
L1947303
1g Each for $84.82

AAL1947306

  SDS
alfa aesar™
L1947306
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1947309

  SDS
alfa aesar™
L1947309
10g Each for $551.47
Description & Specifications

Specifications

MDL Number MFCD03456189
Synonym 4-Carboxypolystyrene
Concentration 1.6 to 2.1mmol/g
Quantity 1g
Chemical Name or Material Benzoic acid on polystyrene