Please login to your online account to display your discounted pricing

Benzoic Acid Copper(II) Salt Hydrate 96%, ACROS Organics™


Quantity 10g
Packaging Glass bottle

Manufacturer: fisher scientific acros  401910100

 View more versions of this product

Catalog No. AC401910100

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Infrared Spectrum Authentic
Loss on Drying 25% max. (1 g, 110°C, 1 hr.)
Packaging Glass bottle
Color Sky-blue
Quantity 10g
Chemical Name or Material Benzoic acid copper(II) salt hydrate
Name Note 96%
CAS 66019-07-6
CAS Min % 95.0
CAS Max % 100.0
Physical Form Sky-blue powder
Assay 95% min.
Assay Percent Range 96%
Molecular Formula C14H10CuO4·xH2O
Linear Formula (C6H5CO2)2Cu·xH2O
Formula Weight 305.77g/mol
MDL Number MFCD03066124
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECS Number 208-552-6
TSCA TSCA

Specifications

Chemical Name or Material Benzoic acid copper(II) salt hydrate
Name Note 96%
CAS 66019-07-6
CAS Min % 95.0
CAS Max % 100.0
Assay 95% min.
Assay Percent Range 96%
Molecular Formula C14H10CuO4·xH2O
Linear Formula (C6H5CO2)2Cu·xH2O
Formula Weight 305.77g/mol
MDL Number MFCD03066124
Accessories

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.

SDS & Safety Documents