Learn More
Benzoic acid, 99%, extra pure, Thermo Scientific™
Benzoic acid, 99%, C7H6O2, CAS Number-65-85-0, benzenecarboxylic acid, benzeneformic acid, benzenemethanoic acid, benzoesaeure gk, benzoesaeure gv, carboxybenzene, dracylic acid, phenylcarboxylic acid, phenylformic acid, retardex, 2.5kg, (20 wt.% soln. in ethanol) clear, 09,92, 249.0 deg.C, 100.0
Supplier: Thermo Scientific Chemicals 149130250
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Benzoic acid | |
98.5 | |
249.0°C | |
Authentic | |
Fibre drum | |
C6H5CO2H | |
25kg | |
01,49; 13,312 | |
benzenecarboxylic acid, dracylic acid, benzeneformic acid, carboxybenzene, phenylformic acid, benzenemethanoic acid, phenylcarboxylic acid, retardex, benzoesaeure gk, benzoesaeure gv | |
WPYMKLBDIGXBTP-UHFFFAOYSA-N | |
benzoic acid | |
243 | |
122.12 | |
(20 wt.% soln. in ethanol) clear |
65-85-0 | |
100.0 | |
121°C | |
99% | |
C7H6O2 | |
MFCD00002398 | |
09,92 | |
15,1093 | |
Solubility in water: 3g/L (20°C). Other solubilities: soluble in alcohol,benzene,ether,chloroform,,ccl4,carbon disulfide and turpentine | |
C1=CC=C(C=C1)C(=O)O | |
122.12 | |
CHEBI:30746 | |
99% | |
Extra Pure |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye damage.
Causes damage to organs through prolonged or repeated exposure if inhaled.
GHS P Statement
Do not breathe dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for s
GHS Signal Word: Danger
EINECSNumber : 200-618-2
RUO â Research Use Only