missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Benzoic acid, 99.6%, for analysis ACS, ACROS Organics™

Manufacturer:  Fisher Scientific Acros 423470250

 View more versions of this product

Catalog No. AC423470250


 

Specifications Description & Specifications

Specifications

≥99.5%
Crystalline Powder
25g
122° to 123°C
0.005% max.
Passes Test
65-85-0
99.5
Plastic bottle
0.005% max. (in methanol)
5ppm max.
ACS Reagent Grade
122.12
White
100.0
C6H5CO2H
Benzoic acid
99.6%
C7H6O2
0.005% max.
Authentic
0.002% max.

Chemical Identifiers

65-85-0
100.0
C7H6O2
122.12
≥99.5%
99.5
99.6%
C6H5CO2H
Benzoic acid
ACS Reagent Grade

Safety and Handling

SDS & Safety Documents

GHS H Statement:
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause an allergic skin reaction.
Harmful if swallowed.
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: If breathing is difficult, remove to fresh air and keep at rest in a position comfortable for breathing.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
Wear eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger