Learn More
Benzoic acid, 99.5%, for analysis, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 221802500
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For analysisSpecifications
Benzoic acid | |
121.0°C to 123.0°C | |
249.0°C | |
100.0 | |
99.5% | |
C6H5CO2H | |
09, 92 | |
15, 1093 | |
Solubility in water: 3g/L (20°C). Other solubilities: soluble in alcohol, benzene, ether, chloroform,, ccl4, carbon disulfide and turpentine | |
C1=CC=C(C=C1)C(=O)O | |
122.123 | |
CHEBI:30746 | |
99.5% | |
250 g | |
Authentic | |
Fine Crystalline Powder |
For analysis | |
65-85-0 | |
99.4 | |
White | |
C7H6O2 | |
MFCD00002398 | |
01,49; 13,312 | |
benzenecarboxylic acid, dracylic acid, benzeneformic acid, carboxybenzene, phenylformic acid, benzenemethanoic acid, phenylcarboxylic acid, retardex, benzoesaeure gk, benzoesaeure gv | |
WPYMKLBDIGXBTP-UHFFFAOYSA-N | |
benzoic acid | |
243 | |
122.12 | |
Analytical | |
121°C | |
Plastic bottle |
Chemical Identifiers
65-85-0 | |
122.123 | |
WPYMKLBDIGXBTP-UHFFFAOYSA-N | |
243 | |
benzoic acid |
C7H6O2 | |
MFCD00002398 | |
benzenecarboxylic acid, dracylic acid, benzeneformic acid, carboxybenzene, phenylformic acid, benzenemethanoic acid, phenylcarboxylic acid, retardex, benzoesaeure gk, benzoesaeure gv | |
CHEBI:30746 | |
C1=CC=C(C=C1)C(=O)O |
Safety and Handling
GHS H Statement:
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause an allergic skin reaction.
Harmful if swallowed.
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: If breathing is difficult, remove to fresh air and keep at rest in a position comfortable for breathing.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
Wear eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guid
Danger
RUO – Research Use Only