Learn More
Benzohydroxamic acid, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1050909
Description
Benzhydroxamic acid is used as precursor in the synthesis of novel mono-anionic and di-anionic hydroxamato complexes by reacting with BiPh3 and Bi(O(t)Bu)3, which has anti-bacterial activity against helicobacter pylori. It is used in the photometric determination of trace amounts of vanadium in alloy steels by making mixed-ligand vanadium chelates with ammonium thiocyanate. It is also involved in the palladium catalyzed synthesis of benzisoxazolones.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsBenzhydroxamic acid is used as precursor in the synthesis of novel mono-anionic and di-anionic hydroxamato complexes by reacting with BiPh3 and Bi(O(t)Bu)3, which has anti-bacterial activity against helicobacter pylori. It is used in the photometric determination of trace amounts of vanadium in alloy steels by making mixed-ligand vanadium chelates with ammonium thiocyanate. It is also involved in the palladium catalyzed synthesis of benzisoxazolones.
Solubility
Soluble in alcohol. Slightly soluble in ether. Insoluble in benzene.
Notes
Store in cool place. Incompatible with strong oxidizing agents.
Specifications
Benzohydroxamic acid | |
∼125°C (decomposition) | |
MFCD00002109 | |
1907585 | |
Soluble in alcohol. Slightly soluble in ether. Insoluble in benzene. | |
C1=CC=C(C=C1)C(=O)NO | |
137.138 | |
137.14 |
495-18-1 | |
C7H7NO2 | |
10 g | |
benzohydroxamic acid, benzhydroxamic acid, benzamide, n-hydroxy, benzohydroxamate, phenylhydroxamic acid, benzohyroxamate, hydroxylamine, n-benzoyl, benzoylhydroxamic acid, n-hydroxy-benzamide, benzenecarbohydroxamic acid | |
VDEUYMSGMPQMIK-UHFFFAOYSA-N | |
N-hydroxybenzamide | |
10313 | |
98% |
Chemical Identifiers
495-18-1 | |
137.138 | |
VDEUYMSGMPQMIK-UHFFFAOYSA-N | |
10313 | |
C1=CC=C(C=C1)C(=O)NO |
C7H7NO2 | |
MFCD00002109 | |
benzohydroxamic acid, benzhydroxamic acid, benzamide, n-hydroxy, benzohydroxamate, phenylhydroxamic acid, benzohyroxamate, hydroxylamine, n-benzoyl, benzoylhydroxamic acid, n-hydroxy-benzamide, benzenecarbohydroxamic acid | |
N-hydroxybenzamide |
Safety and Handling
GHS H Statement
H341-H315-H319-H335
Suspected of causing genetic defects.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P201-P202-P261-P264b-P271-P280i-P281-P302+P352-P304+P340-P305+P351+P338-P308+P313-P332+P313-P362-P501c
H315-H319-H335-H341
EINECSNumber : 207-797-6
RTECSNumber : DF9650000
TSCA : Yes
Recommended Storage : Keep cold
RUO – Research Use Only