missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Benzofuroxan-5-carboxylic acid, 97%

Manufacturer:  Alfa Aesar™ L1201103

 View more versions of this product

Catalog No. AAL1201103



Specifications

Specifications

Benzofuroxan-5-carboxylic acid
1g
C7H4N2O4
612513
AXRCJZJVVZUOHN-UHFFFAOYSA-N
1-oxido-2,1,3-benzoxadiazol-1-ium-5-carboxylic acid
335281
97%
125°C to 131°C
6086-24-4
MFCD00068056
5-carboxy-2,1,3-benzoxadiazol-1-ium-1-olate, benzofuroxan-5-carboxylic acid, 1-hydroxybenzo c 1,2,5-oxadiazole-5-carboxylic acid, 2,1,3-benzoxadiazole-5-carboxylic acid 1-oxide, 2,1,3-benzoxadiazole-5-carboxylic acid n-oxide, 2,1,3-benzoxadiazole-5-carboxylicacid, 1-oxide, 2,1,3-benzoxadiazole-5-carboxylic acid n-oxide, 1-oxo-2,1??,3-benzoxadiazole-5-carboxylic acid
C1=CC2=[N+](ON=C2C=C1C(=O)O)[O-]
180.119
180.12

Chemical Identifiers

6086-24-4
180.119
AXRCJZJVVZUOHN-UHFFFAOYSA-N
335281
C1=CC2=[N+](ON=C2C=C1C(=O)O)[O-]
C7H4N2O4
MFCD00068056
5-carboxy-2,1,3-benzoxadiazol-1-ium-1-olate, benzofuroxan-5-carboxylic acid, 1-hydroxybenzo c 1,2,5-oxadiazole-5-carboxylic acid, 2,1,3-benzoxadiazole-5-carboxylic acid 1-oxide, 2,1,3-benzoxadiazole-5-carboxylic acid n-oxide, 2,1,3-benzoxadiazole-5-carboxylicacid, 1-oxide, 2,1,3-benzoxadiazole-5-carboxylic acid n-oxide, 1-oxo-2,1??,3-benzoxadiazole-5-carboxylic acid
1-oxido-2,1,3-benzoxadiazol-1-ium-5-carboxylic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents