missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Benzofurazan-4-sulfonyl chloride, 97%

Manufacturer:  Alfa Aesar™ H5427503

 View more versions of this product

Catalog No. AAH5427503



Specifications

Specifications

Benzofurazan-4-sulfonyl chloride
1g
C6H3ClN2O3S
UN3261
SIYWVPMYOGMRDW-UHFFFAOYSA-N
2,1,3-benzoxadiazole-4-sulfonyl chloride
2735446
97%
Moisture sensitive
114322-14-4
MFCD00068072
2,1,3-benzoxadiazole-4-sulfonylchloride
C1=CC2=NON=C2C(=C1)S(=O)(=O)Cl
218.611
218.61

Chemical Identifiers

114322-14-4
218.611
SIYWVPMYOGMRDW-UHFFFAOYSA-N
2735446
C1=CC2=NON=C2C(=C1)S(=O)(=O)Cl
C6H3ClN2O3S
MFCD00068072
2,1,3-benzoxadiazole-4-sulfonylchloride
2,1,3-benzoxadiazole-4-sulfonyl chloride
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H335

DOTInformation : Hazard Class: 8; Packaging Group: III

TSCA : No

Recommended Storage : Ambient temperatures

Documents