missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Benzocyclobutene, TCI America™

$55.00 - $181.81

Chemical Identifiers

CAS 694-87-1
Molecular Formula C8H8
Molecular Weight (g/mol) 104.152
MDL Number MFCD01321219
InChI Key UMIVXZPTRXBADB-UHFFFAOYSA-N
Synonym Bicyclo[4.2.0]octa-1,3,5-triene
PubChem CID 69667
ChEBI CHEBI:87328
IUPAC Name bicyclo[4.2.0]octa-1,3,5-triene
SMILES C1CC2=CC=CC=C21
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B38631G View Documents TCI America
B38631G
1g Each for $181.81
 
B3863200MG View Documents TCI America
B3863200MG
200mg Each for $55.00
 
Specifications

Chemical Identifiers

694-87-1
104.152
UMIVXZPTRXBADB-UHFFFAOYSA-N
69667
bicyclo[4.2.0]octa-1,3,5-triene
C8H8
MFCD01321219
Bicyclo[4.2.0]octa-1,3,5-triene
CHEBI:87328
C1CC2=CC=CC=C21

Specifications

150°C
1g
C8H8
3295
UMIVXZPTRXBADB-UHFFFAOYSA-N
bicyclo[4.2.0]octa-1,3,5-triene
69667
104.15
≥94.0% (GC)
Yellow
694-87-1
MFCD01321219
Bicyclo[4.2.0]octa-1,3,5-triene
C1CC2=CC=CC=C21
104.152
CHEBI:87328
Clear Liquid at 20°C
Benzocyclobutene
Safety and Handling

Safety and Handling

TSCA : No